Please login to your online account to display your discounted pricing

CGS 19755, Tocris


Potent, competitive NMDA antagonist

Quantity 10mg

Manufacturer: r & d systems  RDS124110

 View more versions of this product

Catalog No. 124110

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 223.17
Quantity 10mg
Chemical Name or Material CGS 19755
Assay Percent Range >98%
Molecular Formula C7H14NO5P
CAS 110347-85-8
Synonym cis-4-[Phosphomethyl]-piperidine-2-carboxylic acid
Recommended Storage Store at Room Temperature

Specifications

Chemical Name or Material CGS 19755
Assay Percent Range >98%
Molecular Formula C7H14NO5P
Formula Weight 223.17
Synonym cis-4-[Phosphomethyl]-piperidine-2-carboxylic acid
Accessories

Store at Room Temperature