Please login to your online account to display your discounted pricing

CGS 19755, Tocris

Potent, competitive NMDA antagonist

$239.63 - $976.15

Chemical Identifiers

Chemical Name or Material CGS 19755
Assay Percent Range >98%
Molecular Formula C7H14NO5P
Formula Weight 223.17
Synonym cis-4-[Phosphomethyl]-piperidine-2-carboxylic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

124110

 
r & d systems
RDS124110
10mg Each for $239.63

124150

 
r & d systems
RDS124150
50mg Each for $976.15
Description & Specifications

Specifications

Formula Weight 223.17
Quantity 10mg
Molecular Formula C7H14NO5P
Assay Percent Range >98%
Synonym cis-4-[Phosphomethyl]-piperidine-2-carboxylic acid
CAS 110347-85-8
Chemical Name or Material CGS 19755
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature