missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ CGS 15943, 99+%

Potent adenosine receptor antagonist

$132.62 - $461.09

Chemical Identifiers

CAS 104615-18-1
Molecular Formula C13H8ClN5O
Molecular Weight (g/mol) 285.691
MDL Number MFCD01529897
InChI Key MSJODEOZODDVGW-UHFFFAOYSA-N
Synonym 9-chloro-2-2-furyl-1,2,4 triazolo 1,5-c quinazolin-5-imine, 9-chloro-2-furan-2-yl-1,2,4 triazolo 1,5-c quinazolin-5-amine, chembl16687, 9-chloro-2-2-furyl 1,2,4 triazolo 1,5-c quinazolin-5-amine, 1,2,4 triazolo 1,5-c quinazolin-5-amine,9-chloro-2-2-furanyl, 9-chloro-2-2-furanyl-1,2,4 triazolo 1,5-c quinazolin-5-amine, 1,2,4 triazolo 1,5-c quinazolin-5-amine, 9-chloro-2-2-furanyl, 9-chloro-2-2-furanyl-1,2,4 triazolo 1,5-c quinazolin-5-amine, 1,2,4 triazolo 1,5-c quinazolin-5-amine, 9-chloro-2-2-furanyl, acmc-20ddlj
PubChem CID 2690
IUPAC Name 9-chloro-2-(furan-2-yl)-[1,2,4]triazolo[1,5-c]quinazolin-5-amine
SMILES C1=COC(=C1)C2=NN3C(=N2)C4=C(C=CC(=C4)Cl)N=C3N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ61313MA View Documents Alfa Aesar™
J61313MA
10mg Each for $132.62
 
AAJ61313MF View Documents Alfa Aesar™
J61313MF
50mg Each for $461.09
 
Specifications

Chemical Identifiers

104615-18-1
285.691
MSJODEOZODDVGW-UHFFFAOYSA-N
2690
C1=COC(=C1)C2=NN3C(=N2)C4=C(C=CC(=C4)Cl)N=C3N
C13H8ClN5O
MFCD01529897
9-chloro-2-2-furyl-1,2,4 triazolo 1,5-c quinazolin-5-imine, 9-chloro-2-furan-2-yl-1,2,4 triazolo 1,5-c quinazolin-5-amine, chembl16687, 9-chloro-2-2-furyl 1,2,4 triazolo 1,5-c quinazolin-5-amine, 1,2,4 triazolo 1,5-c quinazolin-5-amine,9-chloro-2-2-furanyl, 9-chloro-2-2-furanyl-1,2,4 triazolo 1,5-c quinazolin-5-amine, 1,2,4 triazolo 1,5-c quinazolin-5-amine, 9-chloro-2-2-furanyl, 9-chloro-2-2-furanyl-1,2,4 triazolo 1,5-c quinazolin-5-amine, 1,2,4 triazolo 1,5-c quinazolin-5-amine, 9-chloro-2-2-furanyl, acmc-20ddlj
9-chloro-2-(furan-2-yl)-[1,2,4]triazolo[1,5-c]quinazolin-5-amine

Specifications

White
104615-18-1
MFCD01529897
MSJODEOZODDVGW-UHFFFAOYSA-N
9-chloro-2-(furan-2-yl)-[1,2,4]triazolo[1,5-c]quinazolin-5-amine
2690
Solid
CGS 15943
10mg
C13H8ClN5O
9-chloro-2-2-furyl-1,2,4 triazolo 1,5-c quinazolin-5-imine, 9-chloro-2-furan-2-yl-1,2,4 triazolo 1,5-c quinazolin-5-amine, chembl16687, 9-chloro-2-2-furyl 1,2,4 triazolo 1,5-c quinazolin-5-amine, 1,2,4 triazolo 1,5-c quinazolin-5-amine,9-chloro-2-2-furanyl, 9-chloro-2-2-furanyl-1,2,4 triazolo 1,5-c quinazolin-5-amine, 1,2,4 triazolo 1,5-c quinazolin-5-amine, 9-chloro-2-2-furanyl, 9-chloro-2-2-furanyl-1,2,4 triazolo 1,5-c quinazolin-5-amine, 1,2,4 triazolo 1,5-c quinazolin-5-amine, 9-chloro-2-2-furanyl, acmc-20ddlj
C1=COC(=C1)C2=NN3C(=N2)C4=C(C=CC(=C4)Cl)N=C3N
285.691
285.69
≥99%