Please login to your online account to display your discounted pricing

CGS 12066B dimaleate, Tocris


5-HT1B agonist

Quantity 10mg

Manufacturer: r & d systems  063810

 View more versions of this product

Catalog No. 063810

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 10mg
Formula Weight 566.49
Assay Percent Range >98%
Synonym 7-Trifluoromethyl-4-(4-methyl-1-piperazinyl)pyrrolo[1, 2-a]-quinoxaline dimaleate
Chemical Name or Material CGS 12066B dimaleate
Molecular Formula C17H17F3N4·2C4H4O4
CAS 109028-10-6
Recommended Storage Store at Room Temperature

Specifications

Chemical Name or Material CGS 12066B dimaleate
Assay Percent Range >98%
Molecular Formula C17H17F3N4·2C4H4O4
Formula Weight 566.49
Synonym 7-Trifluoromethyl-4-(4-methyl-1-piperazinyl)pyrrolo[1, 2-a]-quinoxaline dimaleate
Accessories

Store at Room Temperature