Please login to your online account to display your discounted pricing

CGP 55845 hydrochloride, Tocris


Potent, selective GABAB antagonist

Quantity 10mg

Manufacturer: r & d systems  RDS124810

 View more versions of this product

Catalog No. 124810

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 438.71
Quantity 10mg
CAS 149184-22-5
Synonym (2S)-3-[[(1S)-1-(3, 4-Dichlorophenyl)ethyl]amino-2-hydroxypropyl](phenylmethyl)phosphinic acid hydrochloride
Assay Percent Range >98%
Molecular Formula C18H22Cl2NO3P·HCl
Chemical Name or Material CGP 55845 hydrochloride
Recommended Storage Store at Room Temperature

Specifications

Chemical Name or Material CGP 55845 hydrochloride
Assay Percent Range >98%
Molecular Formula C18H22Cl2NO3P·HCl
Formula Weight 438.71
Synonym (2S)-3-[[(1S)-1-(3, 4-Dichlorophenyl)ethyl]amino-2-hydroxypropyl](phenylmethyl)phosphinic acid hydrochloride
Accessories

Store at Room Temperature