Please login to your online account to display your discounted pricing

CGP 52411, Tocris

EGFR inhibitor; Also inhibits Aβ42 fibril formation

$192.67 - $812.12

Chemical Identifiers

Chemical Name or Material CGP 52411
Assay Percent Range >99%
Molecular Formula C20H15N3O2
Formula Weight 329.35
Synonym 5, 6-Bis(phenylamino)-1H-isoindole-1, 3(2H)-dione
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

336050

 
r & d systems
336050
50mg Each for $812.12

336010

 
r & d systems
336010
10mg Each for $192.67
Description & Specifications

Specifications

Formula Weight 329.35
Quantity 50mg
Chemical Name or Material CGP 52411
Assay Percent Range >99%
Molecular Formula C20H15N3O2
CAS 145915-58-8
Synonym 5, 6-Bis(phenylamino)-1H-isoindole-1, 3(2H)-dione
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature