Please login to your online account to display your discounted pricing

CGP 46381, Tocris

Brain penetrant, selective GABAB antagonist

$244.81 - $997.23

Chemical Identifiers

Chemical Name or Material CGP 46381
Assay Percent Range >98%
Molecular Formula C10H22NO2P
Formula Weight 219.26
Synonym (3-Aminopropyl)(cyclohexylmethyl)phosphinic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

124750

 
r & d systems
RDS124750
50mg Each for $997.23

124710

 
r & d systems
RDS124710
10mg Each for $244.81
Description & Specifications

Specifications

Formula Weight 219.26
Quantity 50mg
Chemical Name or Material CGP 46381
Assay Percent Range >98%
Synonym (3-Aminopropyl)(cyclohexylmethyl)phosphinic acid
Recommended Storage Store at Room Temperature
Molecular Formula C10H22NO2P
CAS 123691-14-5
Safety and Handling

Store at Room Temperature