Please login to your online account to display your discounted pricing

CGP 39551, Tocris

Potent, selective and competitive NMDA antagonist

$214.12 - $891.94

Chemical Identifiers

Chemical Name or Material CGP 39551
Assay Percent Range >98%
Molecular Formula C8H16NO5P
Formula Weight 237.19
Synonym (E)-(±)-2-Amino-4-methyl-5-phosphono-3-pentenoic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

140910

 
r & d systems
RDS140910
10mg Each for $214.12

140950

 
r & d systems
RDS140950
50mg Each for $891.94
Description & Specifications

Specifications

Formula Weight 237.19
Quantity 10mg
Molecular Formula C8H16NO5P
Assay Percent Range >98%
Chemical Name or Material CGP 39551
Synonym (E)-(±)-2-Amino-4-methyl-5-phosphono-3-pentenoic acid ethyl ester
CAS 127910-32-1
Recommended Storage Desiccate at +4°C
Safety and Handling

Desiccate at +4°C