Please login to your online account to display your discounted pricing

CGP 20712 dihydrochloride, Tocris

Highly potent and selective β1 antagonist

$252.39 - $1,050.42

Chemical Identifiers

Chemical Name or Material CGP 20712 dihydrochloride
Assay Percent Range >96%
Molecular Formula C23H25F3N4O5·2HCl
Formula Weight 567.39
Synonym 1-[2-((3-Carbamoyl-4-hydroxy)phenoxy)ethylamino]-3-[4-(1-methyl-4-trifluoromethyl-2-imidazolyl)phenoxy]-2-propanol dihydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

102410

 
r & d systems
RDS102410
10mg Each for $252.39

102450

 
r & d systems
RDS102450
50mg Each for $1,050.42
Description & Specifications

Specifications

Formula Weight 567.39
Quantity 10mg
Recommended Storage Desiccate at +4°C
Molecular Formula C23H25F3N4O5·2HCl
CAS 1216905-73-5
Assay Percent Range >96%
Chemical Name or Material CGP 20712 dihydrochloride
Synonym 1-[2-((3-Carbamoyl-4-hydroxy)phenoxy)ethylamino]-3-[4-(1-methyl-4-trifluoromethyl-2-imidazolyl)phenoxy]-2-propanol dihydrochloride
Safety and Handling

Desiccate at +4°C