Please login to your online account to display your discounted pricing

CGP 13501, Tocris

Positive modulator at GABAB receptors

$181.19 - $762.58

Chemical Identifiers

Chemical Name or Material CGP 13501
Assay Percent Range >98%
Molecular Formula C19H30O2
Formula Weight 290.44
Synonym 3,5-bis(1,1-Dimethylethyl)-4-hydroxy-α,α-dimethylbenzenepropanal
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

151410

 
r & d systems
RDS151410
10mg Each for $181.19

151450

 
r & d systems
RDS151450
50mg Each for $762.58
Description & Specifications

Specifications

Formula Weight 290.44
Quantity 10mg
Chemical Name or Material CGP 13501
Assay Percent Range >98%
Molecular Formula C19H30O2
CAS 56189-68-5
Synonym 3,5-bis(1,1-Dimethylethyl)-4-hydroxy-α,α-dimethylbenzenepropanal
Recommended Storage Desiccate at +4°C
Safety and Handling

Desiccate at +4°C