Please login to your online account to display your discounted pricing

CGP 12177 hydrochloride, Tocris

β3 partial agonist. β12 antagonist

$95.70 - $321.56

Chemical Identifiers

Chemical Name or Material CGP 12177 hydrochloride
Assay Percent Range >98%
Molecular Formula C14H21N3O3·HCl
Formula Weight 315.8
Synonym 4-[3-[(1, 1-Dimethylethyl)amino]2-hydroxypropoxy]-1, 3-dihydro-2H-benzimidazol-2-one hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

113410

 
r & d systems
113410
10mg Each for $95.70

113450

 
r & d systems
113450
50mg Each for $321.56
Description & Specifications

Specifications

Formula Weight 315.8
Quantity 10mg
Molecular Formula C14H21N3O3·HCl
Synonym 4-[3-[(1, 1-Dimethylethyl)amino]2-hydroxypropoxy]-1, 3-dihydro-2H-benzimidazol-2-one hydrochloride
Recommended Storage Store at Room Temperature
Assay Percent Range >98%
CAS 64208-32-8
Chemical Name or Material CGP 12177 hydrochloride
Safety and Handling

Store at Room Temperature