Please login to your online account to display your discounted pricing

cGMP Parameter Assay Kit, R&D Systems

$407.05 - $1,953.58

Specifications

Cross Reactivity Cross-species reactivity not tested. <0.5% cross-reactivity observed with available related molecules.
For Use With (Application) ELISA
Sensitivity 3.06pmol/mL
Species Multi-species
View More Specs
Products
Catalog Number Mfr. No. Sufficient For Product Type Quantity Price Quantity    

KGE003

 
r & d systems
RDSKGE003
1 x 96-well plate ELISA kit 1 kit Each for $407.05

SKGE003

 
r & d systems
RDSSKGE003
6 x 96-well plates ELISA kit 1 Pk. Each for $1,953.58
Description & Specifications

Specifications

Cross Reactivity Cross-species reactivity not tested. <0.5% cross-reactivity observed with available related molecules.
For Use With (Application) ELISA
Sensitivity 3.06pmol/mL
Species Multi-species

Assay Type: Solid Phase Competitive ELISA
Sample Type/Volume Required: Cell Culture Supernates (100μL), Cell Lysates (100μL), EDTA Plasma (100μL), Saliva (100μL), Urine (100μL)
Sensitivity: 3.06 pmol/mL
Specificity: Measures cyclic GMP levels in various samples
Cross Reactivity: Cross-species reactivity not tested. <0.5% cross-reactivity observed with available related molecules.