Please login to your online account to display your discounted pricing

cGMP Parameter Assay Kit, R&D Systems


Manufacturer: r & d systems  RDSSKGE003

 View more versions of this product

Catalog No. SKGE003

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) For the quantitative determination of cyclic GMP concentrations in cell culture supernates, plasma, saliva, urine and cell lysates
Product Type ELISA kit
Quantity 1 Pk.
Sensitivity 3.06pmol/mL
Species Multi-species
Sufficient For 6 x 96-well plates
Cross Reactivity Cross-species reactivity not tested. <0.5% cross-reactivity observed with available related molecules.