Please login to your online account to display your discounted pricing

CGK 733, Tocris

Induces apoptosis in prematurely senescent cells

$192.67 - $812.12

Chemical Identifiers

Chemical Name or Material CGK 733
Assay Percent Range >98%
Molecular Formula C23H18Cl3FN4O3S
Formula Weight 555.84
Synonym α-Phenyl-N-[2, 2, 2-trichloro-1-[[[(4-fluoro-3-nitrophenyl)amino]thioxomethyl]amino]ethyl]benzeneacetamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

263950

 
r & d systems
RDS263950
50mg Each for $812.12

263910

 
r & d systems
RDS263910
10mg Each for $192.67
Description & Specifications

Specifications

Formula Weight 555.84
Quantity 50mg
Chemical Name or Material CGK 733
Assay Percent Range >98%
Molecular Formula C23H18Cl3FN4O3S
CAS 905973-89-9
Synonym α-Phenyl-N-[2, 2, 2-trichloro-1-[[[(4-fluoro-3-nitrophenyl)amino]thioxomethyl]amino]ethyl]benzeneacetamide
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C