Please login to your online account to display your discounted pricing

CGH 2466 dihydrochloride, Tocris

A1, A2B and A3 antagonist and inhibitor of p38 MAPK and PDE4

$177.37 - $746.48

Chemical Identifiers

Chemical Name or Material CGH 2466 dihydrochloride
Assay Percent Range >99%
Molecular Formula C14H9N3SCl2·2HCl
Formula Weight 395.13
Synonym 4-(3, 4-Dichlorophenyl)-5-(4-pyridinyl)-2-thiazolamine dihydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

234410

 
r & d systems
RDS234410
10mg Each for $177.37

234450

 
r & d systems
RDS234450
50mg Each for $746.48
Description & Specifications

Specifications

Formula Weight 395.13
Quantity 10mg
Molecular Formula C14H9N3SCl2·2HCl
Chemical Name or Material CGH 2466 dihydrochloride
Synonym 4-(3, 4-Dichlorophenyl)-5-(4-pyridinyl)-2-thiazolamine dihydrochloride
Assay Percent Range >99%
CAS 1177618-54-0
Recommended Storage Desiccate at +4°C
Safety and Handling

Desiccate at +4°C