Please login to your online account to display your discounted pricing

CFTRinh 172, Tocris™

Voltage-independent, selective CFTR chloride channel blocker

$214.12 - $895.76

Chemical Identifiers

Chemical Name or Material CFTRinh 172
Assay Percent Range >98%
Molecular Formula C18H10F3NO3S2
Formula Weight 409.4
Synonym 4-[[4-Oxo-2-thioxo-3-[3-trifluoromethyl)phenyl]-5-thiazolidinylidene]methyl]benzoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

343010

 
r & d systems
RDS343010
10mg Each for $214.12

343050

 
r & d systems
RDS343050
50mg Each for $895.76
Description & Specifications

Specifications

Formula Weight 409.4
Quantity 10mg
Molecular Formula C18H10F3NO3S2
CAS 307510-92-5
Assay Percent Range >98%
Synonym 4-[[4-Oxo-2-thioxo-3-[3-trifluoromethyl)phenyl]-5-thiazolidinylidene]methyl]benzoic acid
Chemical Name or Material CFTRinh 172
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C