Please login to your online account to display your discounted pricing

CFM-2, Tocris


Non-competitive AMPA antagonist

Quantity 50mg

Manufacturer: r & d systems  108250

 View more versions of this product

Catalog No. 108250

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 311.34
Quantity 50mg
Chemical Name or Material CFM-2
Assay Percent Range >98%
Molecular Formula C17H17N3O3
CAS 178616-26-7
Synonym 1-(4'-Aminophenyl)-3,5-dihydro-7,8-dimethoxy-4H-2,3-benzodiazepin-4-one
Recommended Storage Store at Room Temperature

Specifications

Chemical Name or Material CFM-2
Assay Percent Range >98%
Molecular Formula C17H17N3O3
Formula Weight 311.34
Synonym 1-(4'-Aminophenyl)-3,5-dihydro-7,8-dimethoxy-4H-2,3-benzodiazepin-4-one
Accessories

Store at Room Temperature