Please login to your online account to display your discounted pricing

CFM-2, Tocris

Non-competitive AMPA antagonist

$239.63 - $976.15

Chemical Identifiers

Chemical Name or Material CFM-2
Assay Percent Range >98%
Molecular Formula C17H17N3O3
Formula Weight 311.34
Synonym 1-(4'-Aminophenyl)-3,5-dihydro-7,8-dimethoxy-4H-2,3-benzodiazepin-4-one
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

108250

 
r & d systems
108250
50mg Each for $976.15

108210

 
r & d systems
108210
10mg Each for $239.63
Description & Specifications

Specifications

Formula Weight 311.34
Quantity 50mg
CAS 178616-26-7
Recommended Storage Store at Room Temperature
Molecular Formula C17H17N3O3
Assay Percent Range >98%
Synonym 1-(4'-Aminophenyl)-3,5-dihydro-7,8-dimethoxy-4H-2,3-benzodiazepin-4-one
Chemical Name or Material CFM-2
Safety and Handling

Store at Room Temperature