Please login to your online account to display your discounted pricing

Alfa Aesar™ Cesium triiodide, 98%

$28.76 - $28.76

Chemical Identifiers

Chemical Name or Material Cesium triiodide
CAS 20202-54-4
Molecular Formula CsI3
Formula Weight 513.62g/mol
MDL Number MFCD00016053
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1717303

 
alfa aesar™
A17173-03
1g Each for $28.76
Description & Specifications

Specifications

Quantity 1g
Chemical Name or Material Cesium triiodide
CAS 20202-54-4
Assay 98%
Molecular Formula CsI3
Formula Weight 513.62g/mol
MDL Number MFCD00016053
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.