missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cesium sulfate, 99.99% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 1455922

 View more versions of this product

Catalog No. AA1455922



Specifications

Specifications

Cesium sulfate
99.99% (Metal basis)
MFCD00010959
cesium sulfate, dicesium sulfate, sulfuric acid, dicesium salt, caesium sulphate, caesium sulfate, unii-8d6r91cs62, dicaesium 1+ ion sulfate, cesium sulfate, cesiumsulfate, dicaesium 1+ sulfate
[O-]S(=O)(=O)[O-].[Cs+].[Cs+]
361.867
361.87
4.24g/cm3 at 20°C
100g
10294-54-9
Cs2O4S
White powder
FLJPGEWQYJVDPF-UHFFFAOYSA-L
dicesium;sulfate
25137
Powder
White

Chemical Identifiers

10294-54-9
361.867
FLJPGEWQYJVDPF-UHFFFAOYSA-L
25137
[O-]S(=O)(=O)[O-].[Cs+].[Cs+]
Cs2O4S
MFCD00010959
cesium sulfate, dicesium sulfate, sulfuric acid, dicesium salt, caesium sulphate, caesium sulfate, unii-8d6r91cs62, dicaesium 1+ ion sulfate, cesium sulfate, cesiumsulfate, dicaesium 1+ sulfate
dicesium;sulfate
Safety and Handling

Safety and Handling

EINECSNumber : 233-662-6

Documents