Alfa Aesar™ Cesium propionate, complex with propionic acid
Manufacturer:  Alfa Aesar™ 1886509
 View more versions of this product
Catalog No. AA1886509
Specifications
Specifications
Cesium propionate
10g
38869-24-8
MFCD00014025
YBZSHUAKOJGWRT-UHFFFAOYSA-M
cesium;propanoate
23693496
Crystalline
Hygroscopic
Complex with propionic acid
C3H5CsO2
cesium propionate, caesium propanoate, cesium propionate, compound with propionic acid, acmc-20alh6, propionic acid cesium salt, caesium 1+ propionate, caesium 1+ ion propionate
CCC(=O)[O-].[Cs+]
205.976
280.06
Chemical Identifiers
38869-24-8
205.976
YBZSHUAKOJGWRT-UHFFFAOYSA-M
23693496
CCC(=O)[O-].[Cs+]
C3H5CsO2
MFCD00014025
cesium propionate, caesium propanoate, cesium propionate, compound with propionic acid, acmc-20alh6, propionic acid cesium salt, caesium 1+ propionate, caesium 1+ ion propionate
cesium;propanoate
Safety and Handling
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
SDS Documents