missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Cesium perchlorate, Reagent Grade

Manufacturer:  Alfa Aesar™ 1230814

 View more versions of this product

Catalog No. AA1230814



Specifications

Specifications

Cesium perchlorate
ClCsO4
UN1481
WKDKOOITVYKILI-UHFFFAOYSA-M
cesium;perchlorate
123309
Powder
Odorless
250°C (decomposition)
13454-84-7
MFCD00010956
cesium perchlorate, caesium 1+ ion perchlorate, perchloric acid, cesium salt, caesiumperchlorat, caesium perchlorate, perchloric acid, cesium salt 1:1, clo4.cs, acmc-20alh8, cesium perchlorate, caesium 1+ perchlorate ion
[O-]Cl(=O)(=O)=O.[Cs+]
232.351
232.35
Reagent
Hygroscopic
25g

Chemical Identifiers

13454-84-7
232.351
WKDKOOITVYKILI-UHFFFAOYSA-M
123309
[O-]Cl(=O)(=O)=O.[Cs+]
ClCsO4
MFCD00010956
cesium perchlorate, caesium 1+ ion perchlorate, perchloric acid, cesium salt, caesiumperchlorat, caesium perchlorate, perchloric acid, cesium salt 1:1, clo4.cs, acmc-20alh8, cesium perchlorate, caesium 1+ perchlorate ion
cesium;perchlorate
Safety and Handling

Safety and Handling

GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P221-P210-P220-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Keep/Store away from clothing/combustible materials.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: PERCHLORATES, INORGANIC, N.O.S.

TSCA : TSCA

SDS Documents