Please login to your online account to display your discounted pricing

Cesium perchlorate 99% , ACROS Organics™

Chemical Identifiers

Chemical Name or Material Cesium perchlorate
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula ClCsO4
Linear Formula CsClO4
Formula Weight 232.36
MDL Number MFCD00010956
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC389760100

  SDS
thermo fisher scientific
AC389760100
Glass bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC389760500

  SDS
thermo fisher scientific
AC389760500
Glass bottle 50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Crystals or Crystalline Powder
Formula Weight 232.36
Quantity 10g
Color White
Chemical Name or Material Cesium perchlorate
Assay Percent Range 99%
MDL Number MFCD00010956
CAS 13454-84-7
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May intensify fire; oxidizer.
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Molecular Formula ClCsO4
Linear Formula CsClO4
Health Hazard 1 GHS Signal Word: Danger

Oxidation reagent

Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May intensify fire; oxidizer.