Learn More
Cesium nitrate, 99.99%, (trace metal basis), for analysis, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 192070500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysisSpecifications
Cesium nitrate | |
7789-18-6 | |
Crystalline Powder and/or Crystals | |
99.99% | |
CsNO3 | |
15,2014 | |
Solubility in water: 9.16 g/100mL (0°C). Other solubilities: soluble in acetone, slightly soluble in alcohol | |
[N+](=O)([O-])[O-].[Cs+] | |
194.91 | |
194.91 | |
Trace Metal Basis | |
3.68 |
414.0°C | |
White | |
50 g | |
CsNO3 | |
MFCD00010963 | |
cesium nitrate, caesium nitrate, nitric acid, cesium salt, cesium i nitrate 1:1, caesium 1+ nitronate, acmc-1bhhg, caesium 1+ ion nitronate, ksc492a7j | |
NLSCHDZTHVNDCP-UHFFFAOYSA-N | |
cesium;nitrate | |
62674 | |
99.99% | |
Glass bottle | |
3.6800g/mL |
Chemical Identifiers
7789-18-6 | |
194.91 | |
NLSCHDZTHVNDCP-UHFFFAOYSA-N | |
62674 | |
[N+](=O)([O-])[O-].[Cs+] |
CsNO3 | |
MFCD00010963 | |
cesium nitrate, caesium nitrate, nitric acid, cesium salt, cesium i nitrate 1:1, caesium 1+ nitronate, acmc-1bhhg, caesium 1+ ion nitronate, ksc492a7j | |
cesium;nitrate |
Safety and Handling
GHS H Statement
May cause fire or explosion; strong oxidizer.
Harmful if swallowed.
Suspected of damaging fertility or the unborn child.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove conta
GHS Signal Word: Danger
EINECSNumber : 232-146-8
RUO – Research Use Only