Please login to your online account to display your discounted pricing

Cesium nitrate, 99.99% (metals basis), Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Cesium nitrate
CAS 7789-18-6
Molecular Formula CsNO3
Formula Weight 194.91
MDL Number MFCD00010963
UN Number UN1451
Merck Index 14,2015
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA1444036

 
alfa aesar
1444036
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 194.91
Quantity 500g
Chemical Name or Material Cesium nitrate
TSCA TSCA
Odor Odorless
Physical Form Crystalline
Refractive Index 1.55
Assay 99.99%
Sensitivity Hygroscopic
Assay Percent Notes (metals basis)
Molecular Formula CsNO3
MDL Number MFCD00010963
CAS 7789-18-6
UN Number UN1451
Merck Index 14,2015
Color White
Melting Point 414°C
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H272-H315-H319-H335;
May intensify fire; oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P221-P210-P220-P305+P351+P338-P405-P501a;
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Keep/Store away from clothing/combustible materials.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Density 3.68
DOT Information Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: CESIUM NITRATE
RTECS Number FL0700000
Safety and Handling

Danger

GHS P Statement
P221-P210-P220-P305+P351+P338-P405-P501a;
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Keep/Store away from clothing/combustible materials.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H272-H315-H319-H335;
May intensify fire; oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.