Learn More
Cesium nitrate, 99.99% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01444036
Description
Cesium nitrate prisms are used in infrared spectroscopy, x-ray phosphors and scintillation counters. It is a precursor material for the synthesis of cesium nitratocuprate and cesium salts. Cesium nitrate doped on carbon-based ruthenium catalyst plays an important role in synthesis of ammonia.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
(decomposition) | |
414°C | |
White | |
500 g | |
CsNO3 | |
UN1451 | |
cesium nitrate, caesium nitrate, nitric acid, cesium salt, cesium i nitrate 1:1, caesium 1+ nitronate, acmc-1bhhg, caesium 1+ ion nitronate, ksc492a7j | |
NLSCHDZTHVNDCP-UHFFFAOYSA-N | |
cesium;nitrate | |
62674 | |
99.99% | |
1.55 | |
3.68 |
Cesium nitrate | |
7789-18-6 | |
Crystalline | |
(metals basis) | |
MFCD00010963 | |
14,2015 | |
Soluble in water and acetone. Slightly soluble in ethanol. | |
[N+](=O)([O-])[O-].[Cs+] | |
194.909 | |
194.91 | |
Odorless | |
Hygroscopic |
Chemical Identifiers
7789-18-6 | |
194.909 | |
NLSCHDZTHVNDCP-UHFFFAOYSA-N | |
62674 | |
[N+](=O)([O-])[O-].[Cs+] |
CsNO3 | |
MFCD00010963 | |
cesium nitrate, caesium nitrate, nitric acid, cesium salt, cesium i nitrate 1:1, caesium 1+ nitronate, acmc-1bhhg, caesium 1+ ion nitronate, ksc492a7j | |
cesium;nitrate |
Safety and Handling
GHS H Statement
H272-H315-H319-H335;
May intensify fire; oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P201-P202-P220-P221-P264-P270-P272-P280-P301+P312-P308+P313-P330-P370+P378q-P370+P380+P375-P372-P501c
H271-H302-H361f
DOTInformation : Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: CESIUM NITRATE
EINECSNumber : 232-146-8
RTECSNumber : FL0700000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only