missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cesium nitrate, 99.99% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 1444006

 View more versions of this product

Catalog No. AA1444006



Specifications

Specifications

Cesium nitrate
99.99% (Metal basis)
MFCD00010963
cesium nitrate, caesium nitrate, nitric acid, cesium salt, cesium i nitrate 1:1, cesium nitrate, caesium 1+ nitronate, cesium nitrate, acmc-1bhhg, caesium 1+ ion nitronate, ksc492a7j
[N+](=O)([O-])[O-].[Cs+]
194.909
194.91
Odorless
5g
7789-18-6
CsNO3
White crystalline powder
NLSCHDZTHVNDCP-UHFFFAOYSA-N
cesium;nitrate
62674
Crystalline
White

Chemical Identifiers

7789-18-6
194.909
NLSCHDZTHVNDCP-UHFFFAOYSA-N
62674
[N+](=O)([O-])[O-].[Cs+]
CsNO3
MFCD00010963
cesium nitrate, caesium nitrate, nitric acid, cesium salt, cesium i nitrate 1:1, cesium nitrate, caesium 1+ nitronate, cesium nitrate, acmc-1bhhg, caesium 1+ ion nitronate, ksc492a7j
cesium;nitrate
Safety and Handling

Safety and Handling

H272-H315-H319-H335-H303 : May intensify fire; oxidizer. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. May be harmful if swallowed.

EINECSNumber : 232-146-8

SDS Documents