Please login to your online account to display your discounted pricing

Alfa Aesar™ Cesium nitrate, 99.9% (metals basis)

Chemical Identifiers

Chemical Name or Material Cesium nitrate
CAS 7789-18-6
Formula Weight 194.91g/mol
UN Number UN1451
Merck Index 14,2015
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1002306

 
alfa aesar™
10023-06
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Density 3.68g/mL
Mesh Size −20 Mesh
Odor Odorless
Quantity 5g
Refractive Index 1.55
Boiling Point (dec.)
Color White
Melting Point 414°C
Quantity 5g
Chemical Name or Material Cesium nitrate
CAS 7789-18-6
Physical Form Powder
Assay 99.9%
Assay Percent Notes (metals basis)
Formula Weight 194.91g/mol
UN Number UN1451
Merck Index 14,2015
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P221-P210-P220-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Keep/Store away from clothing/combustible materials.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: CESIUM NITRATE
RTECS Number FL0700000
TSCA TSCA
Safety and Handling

Danger

GHS P Statement
P221-P210-P220-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Keep/Store away from clothing/combustible materials.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.