Please login to your online account to display your discounted pricing

Alfa Aesar™ Cesium iodide, ultra dry, 99.998% (metals basis)

$116.47 - $116.47

Chemical Identifiers

Chemical Name or Material Cesium iodide
CAS 7789-17-5
Name Note Ultra dry
Formula Weight 259.81
MDL Number MFCD00010962
Merck Index 14,2014
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3572903

  SDS
alfa aesar™
3572903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3572909

  SDS
alfa aesar™
3572909
10g Each for $116.47

AA3572918

  SDS
alfa aesar™
3572918
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3572930

  SDS
alfa aesar™
3572930
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 259.81
Packaging Ampule
Health Hazard 2 GHS H Statement
H302-H315-H319-H317-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Melting Point 626°C
Storage Note 1
  • Ampouled under argon
Assay Percent Notes (metals basis)
Assay 99.998%
Health Hazard 1 Warning
CAS 7789-17-5
TSCA TSCA
Name Note Ultra dry
Refractive Index 1.7876
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 1280°C
Mesh Size −10 Mesh
Color White
MDL Number MFCD00010962
Sensitivity Hygroscopic
Merck Index 14,2014
Quantity 1g
Physical Form Beads
Odor Odorless
Chemical Name or Material Cesium iodide

  • Ampouled under argon

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H315-H319-H317-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.

  • Ampouled under argon