Learn More
Cesium iodide, 99.999%, (trace metal basis), Thermo Scientific™
Supplier: Thermo Scientific Chemicals 192820500
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
For ultra trace analysisSpecifications
Cesium iodide | |
50g | |
CsI | |
15,2013 | |
Solubility in water: 740g/L (20°C). Other solubilities: soluble in ethanol,slightly soluble in methanol,practically insoluble in acetone | |
[I-].[Cs+] | |
259.8 | |
259.8 | |
Trace Metal Basis |
7789-17-5 | |
99.999% | |
CsI | |
cesium iodide, caesium iodide, cesium monoiodide, dicesium diiodide, tricesium triiodide, cesiumiodide, cesium iodide cs2i2, cesium iodide cs3i3, unii-u1p3gvc56l | |
XQPRBTXUXXVTKB-UHFFFAOYSA-M | |
cesium;iodide | |
24601 | |
99.999% | |
Glass bottle |
Safety and Handling
GHS H Statement
Suspected of damaging fertility or the unborn child if inhaled.
Very toxic to aquatic life.
GHS P Statement
Obtain special instructions before use.
Use personal protective equipment as required.
IF exposed or concerned: Get medical advice/attention.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the
GHS Signal Word: Warning
EINECSNumber : 232-145-2
RUO â Research Use Only