Learn More
Cesium iodide, 99.999% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01099214
Description
Cesium iodide is used as an input phosphor in x-ray image intensifier tube of fluoroscopy equipment. It is also used as an equipment for electromagnetic calorimetry in experimental particle physics. It is often employed as a beamsplitter in Fourier Transform Infrared (FT-IR) spectrometers. Cesium iodide is an inorganic scintillator which is mainly used in experimental nuclear physics, high energy physics, nuclear medicine, nuclear tomography, environmental studies and many other fields of use. It is utilized in calorimeters and detectors and also employed as a phosphor in fluoroscopy equipments.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1280°C | |
626°C | |
White | |
25 g | |
CsI | |
14,2014 | |
Soluble in water. Soluble in water and ethanol. Slightly soluble in methanol. Insoluble in acetone. | |
[I-].[Cs+] | |
259.81 | |
259.81 | |
Odorless | |
Hygroscopic |
Cesium iodide | |
7789-17-5 | |
Crystalline | |
(metals basis) | |
MFCD00010962 | |
cesium iodide, caesium iodide, cesium monoiodide, dicesium diiodide, tricesium triiodide, cesiumiodide, cesium iodide cs2i2, cesium iodide cs3i3, unii-u1p3gvc56l | |
XQPRBTXUXXVTKB-UHFFFAOYSA-M | |
cesium;iodide | |
24601 | |
99.999% | |
1.7876 |
Chemical Identifiers
7789-17-5 | |
259.81 | |
XQPRBTXUXXVTKB-UHFFFAOYSA-M | |
24601 | |
[I-].[Cs+] |
CsI | |
MFCD00010962 | |
cesium iodide, caesium iodide, cesium monoiodide, dicesium diiodide, tricesium triiodide, cesiumiodide, cesium iodide cs2i2, cesium iodide cs3i3, unii-u1p3gvc56l | |
cesium;iodide |
Safety and Handling
GHS H Statement
H302-H315-H319-H317-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
P201-P202-P261-P264b-P271-P272-P280-P302+P352-P304+P340-P305+P351+P338-P308+P313-P333+P313-P362-P501c
H315-H317-H319-H335-H361
DOTInformation : Hazard Class: 9; Packaging Group: III
EINECSNumber : 232-145-2
RTECSNumber : FLO350000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only