missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Cesium iodide, 99.999% (metals basis)

Manufacturer:  Alfa Aesar™ 1099222

 View more versions of this product

Catalog No. AA1099222



Specifications

Specifications

Cesium iodide
(metals basis)
MFCD00010962
caesium iodide
[I-].[Cs+]
259.81
259.81
99.999%
1.7876
1280°C
626°C
7789-17-5
CsI
14,2014
XQPRBTXUXXVTKB-UHFFFAOYSA-M
cesium;iodide
24601
Crystalline
Odorless
Hygroscopic
White
100g

Chemical Identifiers

7789-17-5
259.81
XQPRBTXUXXVTKB-UHFFFAOYSA-M
24601
[I-].[Cs+]
CsI
MFCD00010962
caesium iodide
cesium;iodide
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H317-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : FLO350000

TSCA : TSCA

Documents