missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cesium ICP Standard, 1mL = 10mg Cs (10,000ppm Cs), Ricca Chemical

Manufacturer:  Ricca Chemical Company RIPCS10KN500

 View more versions of this product

Catalog No. PCS10KN-500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Cesium ICP Standard
90.0,6.76,1.32
CsNO3
[N+](=O)([O-])[O-].[Cs+]
194.909
Liquid
10,000ppm Cs
Colorless
7789-18-6,7697-37-2,7789-18-6
94.0,7.03,1.37
NLSCHDZTHVNDCP-UHFFFAOYSA-N
cesium;nitrate
62674
Poly natural
1 mL = 10 mg Cs
500mL

Chemical Identifiers

7789-18-6
194.909
62674
[N+](=O)([O-])[O-].[Cs+]
CsNO3
NLSCHDZTHVNDCP-UHFFFAOYSA-N
cesium;nitrate
Documents