missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cesium hydroxide monohydrate, 96%; cesium carbonate <5%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2248014

 View more versions of this product

Catalog No. AAB2248014


Description

Description

Cesium hydroxide monohydrate can be used as a strong electrolyte for the battery and it also can be used as polymerization catalyst. It is also used as a raw materials for manufacturing various cesium salts and Catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cesium hydroxide monohydrate can be used as a strong electrolyte for the battery and it also can be used as polymerization catalyst. It is also used as a raw materials for manufacturing various cesium salts and Catalyst.

Solubility
Soluble in water with exothermic reaction.

Notes
Air sensitive & hygroscopic. Incompatible with oxidizing agents, acids, air, water/moisture.
Specifications

Specifications

Cesium hydroxide monohydrate
35103-79-8
cesium carbonate <5%
MFCD00149664
14,2013
Soluble in water with exothermic reaction.
O.[OH-].[Cs+]
167.93
167.93 (149.91 Anhydrous)
Odorless
1.87
205°C to 206°C
25 g
CsH3O2
UN3262
cesium hydroxide monohydrate, cesium hydroxide hydrate, cesium hydroxide, monohydrate, cesium hydroxide-hydrate, cesium hydroxide, hydrate, ksc493c5l, caesium 1+ ion hydrate hydroxide, cesium hydroxide,monohydrate 9ci
ABSOMGPQFXJESQ-UHFFFAOYSA-M
caesium(1+) hydrate hydroxide
23679066
96%
Air sensitive; Hygroscopic

Chemical Identifiers

35103-79-8
167.93
ABSOMGPQFXJESQ-UHFFFAOYSA-M
23679066
CsH3O2
MFCD00149664
cesium hydroxide monohydrate, cesium hydroxide hydrate, cesium hydroxide, monohydrate, cesium hydroxide-hydrate, cesium hydroxide, hydrate, ksc493c5l, caesium 1+ ion hydrate hydroxide, cesium hydroxide,monohydrate 9ci
O.[OH-].[Cs+]
Safety and Handling

Safety and Handling

GHS H Statement
H314-H302
Causes severe skin burns and eye damage.
Harmful if swallowed.

P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302-H314

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, BASIC, INORGANIC, N.O.S.

EINECSNumber : 244-344-1

RTECSNumber : FK9800000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only