missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cesium hydroxide hydrate, 99.9% (metals basis), Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 01323322

 View more versions of this product

Catalog No. AA1323322



Description

Description

Cesium hydroxide hydrate is used as a catalyst for the conversion of alcohols substituted anilines and heterocyclic amines undergo an addition in NMP to phenylacetylene leading to functionalized enol ethers and enamines. It is also used in the chemoselective N-alkylation of secondary amines with alkyl halides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Cesium hydroxide hydrate
White
100 g
CsHO
cesium hydroxide monohydrate, cesium hydroxide hydrate, cesium hydroxide, monohydrate, cesium hydroxide-hydrate, cesium hydroxide, hydrate, ksc493c5l, caesium 1+ ion hydrate hydroxide, cesium hydroxide,monohydrate 9ci
HUCVOHYBFXVBRW-UHFFFAOYSA-M
cesium;hydroxide;hydrate
23679066
Odorless
White crystalline
12260-45-6
Crystalline
99.9% (Metal basis)
MFCD00010964
Soluble in water and ethanol.
[OH-].[Cs+]
149.91
149.91 (Anhydrous)
Hygroscopic

Chemical Identifiers

12260-45-6
149.91
HUCVOHYBFXVBRW-UHFFFAOYSA-M
23679066
CsHO
MFCD00010964
cesium hydroxide monohydrate, cesium hydroxide hydrate, cesium hydroxide, monohydrate, cesium hydroxide-hydrate, cesium hydroxide, hydrate, ksc493c5l, caesium 1+ ion hydrate hydroxide, cesium hydroxide,monohydrate 9ci
[OH-].[Cs+]
Safety and Handling

Safety and Handling

P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302-H314-H335

DOTInformation : DOT Class: 8, Packaging Group: II

EINECSNumber : 244-344-1

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only