missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cesium hydroxide hydrate, 99.9% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 1323306

 View more versions of this product

Catalog No. AA1323306



Specifications

Specifications

Cesium hydroxide hydrate
99.9% (Metal basis)
MFCD00010964
cesium hydroxide monohydrate, cesium hydroxide hydrate, cesium hydroxide, monohydrate, cesium hydroxide-hydrate, cesium hydroxide, hydrate, ksc493c5l, caesium 1+ ion hydrate hydroxide, cesium hydroxide hydrate, cesium hydroxide,monohydrate 9ci
O.[OH-].[Cs+]
167.927
149.91 (Anhydrous)
Odorless
5g
12260-45-6
CsH3O2
White crystalline
ABSOMGPQFXJESQ-UHFFFAOYSA-M
cesium;hydroxide;hydrate
23679066
Crystalline
White

Chemical Identifiers

12260-45-6
167.927
ABSOMGPQFXJESQ-UHFFFAOYSA-M
23679066
O.[OH-].[Cs+]
CsH3O2
MFCD00010964
cesium hydroxide monohydrate, cesium hydroxide hydrate, cesium hydroxide, monohydrate, cesium hydroxide-hydrate, cesium hydroxide, hydrate, ksc493c5l, caesium 1+ ion hydrate hydroxide, cesium hydroxide hydrate, cesium hydroxide,monohydrate 9ci
cesium;hydroxide;hydrate
Safety and Handling

Safety and Handling

H314-H318-H302 :
Causes severe skin burns and eye damage. Causes serious eye damage. Harmful if swallowed.

DOTInformation : DOT Class: 8, Packaging Group: II

EINECSNumber : 244-344-1

Documents