missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Cesium hydroxide, 50% w/w aq. soln., 99% (metals basis)

Manufacturer:  Alfa Aesar™ 16505NF

 View more versions of this product

Catalog No. AA16505NF



Specifications

Specifications

Cesium hydroxide
(metals basis)
MFCD00010964
cesium hydroxide, caesium hydroxide, cesium hydrate, csoh, cesium hydroxide cs oh, cesium hydroxide dimer, cesium hydoxide, cs.ho, caesium 1+ oh, cesium hydroxide solution
[OH-].[Cs+]
149.912
CHEBI:33988
Liquid
50% w/w aq. soln.
Odorless
Colorless
21351-79-1
CsHO
UN2681
HUCVOHYBFXVBRW-UHFFFAOYSA-M
cesium;hydroxide
62750
149.91
99%
1.725g/mL
Air Sensitive
100g

Chemical Identifiers

21351-79-1
149.912
HUCVOHYBFXVBRW-UHFFFAOYSA-M
62750
cesium;hydroxide
CsHO
MFCD00010964
cesium hydroxide, caesium hydroxide, cesium hydrate, csoh, cesium hydroxide cs oh, cesium hydroxide dimer, cesium hydoxide, cs.ho, caesium 1+ oh, cesium hydroxide solution
CHEBI:33988
[OH-].[Cs+]
Safety and Handling

Safety and Handling

GHS H Statement
H314-H302
Causes severe skin burns and eye damage.
Harmful if swallowed.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CAESIUM HYDROXIDE SOLUTION

TSCA : TSCA

SDS Documents