Learn More
Cesium hydroxide, 50% w/w aq. soln., 99.9% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 010020NF
Description
Cesium hydroxide is used in electric storage batteries. Also used to make porous cesium titanosilicates and potential ion-exchange materials for cleanup of radioactive wastes. Cesium Hydroxide is used as electrolyte in alkaline batteries at subzero temperatures, color photography and as a catalyst in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Cesium hydroxide | |
Colorless | |
100 g | |
CsHO | |
UN2681 | |
Highly soluble in water. | |
[OH-].[Cs+] | |
149.91 | |
CHEBI:33988 | |
99.9% | |
Odorless | |
1.725 g/mL |
21351-79-1 | |
Liquid | |
(metals basis) | |
MFCD00010964 | |
cesium hydroxide, caesium hydroxide, cesium hydrate, csoh, cesium hydroxide cs oh, cesium hydroxide dimer, cesium hydoxide, cs.ho, caesium 1+ oh, cesium hydroxide solution | |
HUCVOHYBFXVBRW-UHFFFAOYSA-M | |
cesium;hydroxide | |
62750 | |
149.91 | |
50% w/w aq. soln. | |
Air Sensitive |
Chemical Identifiers
21351-79-1 | |
149.91 | |
HUCVOHYBFXVBRW-UHFFFAOYSA-M | |
62750 | |
cesium;hydroxide |
CsHO | |
MFCD00010964 | |
cesium hydroxide, caesium hydroxide, cesium hydrate, csoh, cesium hydroxide cs oh, cesium hydroxide dimer, cesium hydoxide, cs.ho, caesium 1+ oh, cesium hydroxide solution | |
CHEBI:33988 | |
[OH-].[Cs+] |
Safety and Handling
GHS H Statement
H314-H302
Causes severe skin burns and eye damage.
Harmful if swallowed.
P201-P202-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302-H314-H335-H361
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CAESIUM HYDROXIDE SOLUTION
EINECSNumber : 244-344-1
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only