missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Cesium hydrogen carbonate, 99.99% (metals basis)

Manufacturer:  Alfa Aesar™ 1423414

 View more versions of this product

Catalog No. AA1423414



Specifications

Specifications

Cesium hydrogen carbonate
(metals basis)
MFCD00010961
ZMCUDHNSHCRDBT-UHFFFAOYSA-M
cesium;hydrogen carbonate
2734983
Crystalline Aggregates
Hygroscopic
25g
29703-01-3
CHCsO3
cesium bicarbonate, cesium hydrogen carbonate, caesium bicarbonate, caesium 1+ ion bicarbonate, cesium hydrogencarbonate, acmc-1cmhe, cho3.cs, caesium 1+ bicarbonate, cesium hydrogencarbonate, carbonic acid, cesiumsalt 1:1
C(=O)(O)[O-].[Cs+]
193.921
193.92
99.99%
175°C (decomposition)

Chemical Identifiers

29703-01-3
193.921
ZMCUDHNSHCRDBT-UHFFFAOYSA-M
2734983
C(=O)(O)[O-].[Cs+]
CHCsO3
MFCD00010961
cesium bicarbonate, cesium hydrogen carbonate, caesium bicarbonate, caesium 1+ ion bicarbonate, cesium hydrogencarbonate, acmc-1cmhe, cho3.cs, caesium 1+ bicarbonate, cesium hydrogencarbonate, carbonic acid, cesiumsalt 1:1
cesium;hydrogen carbonate
Documents