missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Cesium Green 85.0+%, TCI America™
Supplier: TCI America C2806250MG
Specifications
Cesium Green | |
C32H33NO8 | |
HCDBZEWQACQBAN-UHFFFAOYSA-N | |
4-[4-[4-[2-[2-[2-[2-(4-nitrophenoxy)ethoxy]ethoxy]ethoxy]ethoxy]phenyl]phenyl]phenol | |
124523977 | |
≥85.0% (HPLC) | |
Crystal/Powder |
1611477-45-2 | |
4′′-[2-[2-[2-[2-(4-Nitrophenoxy)ethoxy]ethoxy]ethoxy]ethoxy]-[1,1′:4′,1′′-terphenyl]-4-ol | |
C1=CC(=CC=C1C2=CC=C(C=C2)O)C3=CC=C(C=C3)OCCOCCOCCOCCOC4=CC=C(C=C4)[N+](=O)[O-] | |
559.615 | |
559.62 | |
250 mg |
Chemical Identifiers
1611477-45-2 | |
559.615 | |
124523977 | |
C1=CC(=CC=C1C2=CC=C(C=C2)O)C3=CC=C(C=C3)OCCOCCOCCOCCOC4=CC=C(C=C4)[N+](=O)[O-] |
C32H33NO8 | |
HCDBZEWQACQBAN-UHFFFAOYSA-N | |
4-[4-[4-[2-[2-[2-[2-(4-nitrophenoxy)ethoxy]ethoxy]ethoxy]ethoxy]phenyl]phenyl]phenol |
Safety and Handling
TSCA : No