Please login to your online account to display your discounted pricing

Cesium formate monohydrate, for analysis, ACROS Organics™


Quantity 100g
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  AC392801000

 View more versions of this product

Catalog No. AC392801000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥99.5%
Physical Form Crystalline Powder
Formula Weight 195.94
Quantity 100g
Color White
Purity Grade For analysis
Calcium (Ca) 0.001% max.
Potassium (K) 0.03% max.
Sulfate 0.01% max.
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Sodium (Na) 0.02% max.
Barium (Ba) 0.005% max.
Chemical Name or Material Cesium formate monohydrate
MDL Number MFCD00039103
Chloride 0.03% max.
Lithium (Li) 0.001% max.
Magnesium (Mg) 0.0002% max.
Strontium (Sr) 0.0005% max.
Packaging Plastic bottle
Rubidium (Rb) 0.005% max.
Linear Formula HCOOCs
Molecular Formula CHCsO2·H2O
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.

Specifications

Chemical Name or Material Cesium formate monohydrate
Purity Grade For analysis
Assay ≥99.5%
Molecular Formula CHCsO2·H2O
Linear Formula HCOOCs
Formula Weight 195.94
MDL Number MFCD00039103
Accessories

GHS P Statement:
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.

SDS & Safety Documents