Please login to your online account to display your discounted pricing

Cesium formate monohydrate, for analysis, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Cesium formate monohydrate
Purity Grade For analysis
Assay ≥99.5%
Molecular Formula CHCsO2·H2O
Linear Formula HCOOCs
Formula Weight 195.94
MDL Number MFCD00039103
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC392801000

  SDS
thermo fisher scientific
AC392801000
Plastic bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC392800250

  SDS
thermo fisher scientific
AC392800250
Plastic bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥99.5%
Physical Form Crystalline Powder
Formula Weight 195.94
Quantity 100g
Color White
Purity Grade For analysis
Chemical Name or Material Cesium formate monohydrate
Infrared Spectrum Authentic
Potassium (K) 0.03% max.
Sodium (Na) 0.02% max.
Sulfate 0.01% max.
Barium (Ba) 0.005% max.
Calcium (Ca) 0.001% max.
Health Hazard 3 GHS P Statement:
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Lithium (Li) 0.001% max.
Magnesium (Mg) 0.0002% max.
Packaging Plastic bottle
Strontium (Sr) 0.0005% max.
MDL Number MFCD00039103
Chloride 0.03% max.
Rubidium (Rb) 0.005% max.
Molecular Formula CHCsO2·H2O
Linear Formula HCOOCs
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Safety and Handling

GHS P Statement:
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.