Please login to your online account to display your discounted pricing

Cesium formate hydrate, White chunks/powder, 99.9% (Metals basis), Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Cesium formate hydrate
Physical Form Chunks and Powder
Assay Percent Range 99.9% (Metals basis)
Molecular Formula CsOOCH·xH2O
Formula Weight 177.92
MDL Number MFCD00039103
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1292722

  SDS
alfa aesar
1292722
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1292714

  SDS
alfa aesar
1292714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1292736

  SDS
alfa aesar
1292736
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Chunks and Powder
Formula Weight 177.92
Quantity 100g
Color White
Chemical Name or Material Cesium formate hydrate
EINECS Number 222-492-8
Assay Percent Range 99.9% (Metals basis)
Molecular Formula CsOOCH·xH2O
MDL Number MFCD00039103
Anhydrous or Hydrate Hydrate
Health Hazard 1 H315-H319 : Causes skin irritation. Causes serious eye irritation.
Health Hazard 2 In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves.
Safety and Handling

H315-H319 : Causes skin irritation. Causes serious eye irritation.

In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves.