Learn More
Cesium fluoride, 99.9% (metals basis), Thermo Scientific Chemicals
$46.30 - $320.38
Chemical Identifiers
CAS | 13400-13-0 |
---|---|
Molecular Formula | CsF |
Molecular Weight (g/mol) | 151.90 |
MDL Number | MFCD00010960 |
InChI Key | XJHCXCQVJFPJIK-UHFFFAOYSA-M |
Synonym | cesium fluoride, caesium fluoride, cesium monofluoride, dicesium difluoride, tricesium trifluoride, unii-t76a371hjr, caesium 1+ ion fluoride, caesiumfluoride, cesiumfluoride |
PubChem CID | 25953 |
SMILES | [F-].[Cs+] |
Description
Used for desilylation reactions in organic synthesis. Useful base in organic chemistry, due to its low nucleophilicity. Owing to its high ionic character, it is a very reactive source of fluoride ion in organic synthesis. Reagent of choice when reactive naked fluoride is required. Effective catalyst in promoting Knoevenegal condensation. Single crystals of CsF are useful as the windows of cells for infrared spectroscopy.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13400-13-0 | |
151.90 | |
XJHCXCQVJFPJIK-UHFFFAOYSA-M | |
25953 |
CsF | |
MFCD00010960 | |
cesium fluoride, caesium fluoride, cesium monofluoride, dicesium difluoride, tricesium trifluoride, unii-t76a371hjr, caesium 1+ ion fluoride, caesiumfluoride, cesiumfluoride | |
[F-].[Cs+] |
Specifications
1251°C | |
13400-13-0 | |
Lump/Powder | |
99.9% (Metals basis) | |
MFCD00010960 | |
Soluble in water. Insoluble in dioxane and pyridine. | |
[F-].[Cs+] | |
151.90 | |
151.9 | |
Cesium fluoride |
682°C | |
White | |
2.5 g | |
CsF | |
cesium fluoride, caesium fluoride, cesium monofluoride, dicesium difluoride, tricesium trifluoride, unii-t76a371hjr, caesium 1+ ion fluoride, caesiumfluoride, cesiumfluoride | |
XJHCXCQVJFPJIK-UHFFFAOYSA-M | |
cesium;fluoride | |
25953 | |
Hygroscopic |
Safety and Handling
P201-P202-P261-P264b-P270-P271-P281-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P310-P311-P312-P330-P332+P313-P361-P363-P501c
H301+H311+H331-H315-H318-H361
DOTInformation : DOT Class: 6.1; Packaging Group: II
EINECSNumber : 236-487-3
RTECSNumber : FK9650000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only