missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cesium fluoride, 99% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 1288514

 View more versions of this product

Catalog No. AA1288514



Specifications

Specifications

Cesium fluoride
99% (Metals basis)
CsF
UN2923
XJHCXCQVJFPJIK-UHFFFAOYSA-M
cesium;fluoride
25953
Crystalline
4.115g/mL
Hygroscopic
White
6.5 to 7.5
13400-13-0
(metals basis)
MFCD00010960
cesium fluoride, caesium fluoride, cesium monofluoride, cesium fluoride, dicesium difluoride, tricesium trifluoride, unii-t76a371hjr, caesium 1+ ion fluoride, caesiumfluoride, cesiumfluoride
[F-].[Cs+]
151.904
151.9
99%
Odorless
1251°C
682°C
25g

Chemical Identifiers

13400-13-0
151.904
XJHCXCQVJFPJIK-UHFFFAOYSA-M
25953
[F-].[Cs+]
CsF
MFCD00010960
cesium fluoride, caesium fluoride, cesium monofluoride, cesium fluoride, dicesium difluoride, tricesium trifluoride, unii-t76a371hjr, caesium 1+ ion fluoride, caesiumfluoride, cesiumfluoride
cesium;fluoride
Safety and Handling

Safety and Handling

GHS H Statement
H300-H310-H330-H314-H318
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.

RTECSNumber : FK9650000

TSCA : TSCA

Documents