Please login to your online account to display your discounted pricing

Cesium fluoride, 99.9%, extra pure, ACROS Organics™


Quantity 100g
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  AC315911000

 View more versions of this product

Catalog No. AC315911000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Purity Grade Extra Pure
Physical Form Powder
Formula Weight 151.91
Quantity 100g
Color White
Fieser 01,121; 06,100; 07,57; 08,81; 09,100; 10,81; 11,115; 12,108; 13,68; 14,79; 15,75; 16,69; 17,68
Merck Index 15, 2011
Chemical Name or Material Cesium fluoride
Packaging Plastic bottle
Molecular Formula CsF
CAS 13400-13-0
Health Hazard 1 Danger
Assay Percent Range 99.9%
Linear Formula CsF
Total Trace Metal Impurities 0.1% max.
Health Hazard 2 GHS P Statement
Toxic in contact with skin.
Causes serious eye damage.
Causes skin irritation.
Toxic if inhaled.
Toxic if swallowed.
Contact with acids liberates very toxic gas.
MDL Number MFCD00010960
Health Hazard 3 GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Cesium fluoride
CAS 13400-13-0
Purity Grade Extra Pure
Assay Percent Range 99.9%
Molecular Formula CsF
Linear Formula CsF
Formula Weight 151.91
MDL Number MFCD00010960
Accessories

Danger

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS P Statement
Toxic in contact with skin.
Causes serious eye damage.
Causes skin irritation.
Toxic if inhaled.
Toxic if swallowed.
Contact with acids liberates very toxic gas.

SDS & Safety Documents