Please login to your online account to display your discounted pricing

Alfa Aesar™ Cesium dihydrogen arsenate, 99.99%

Chemical Identifiers

Chemical Name or Material Cesium dihydrogen arsenate
CAS 16331-85-4
Molecular Formula H2AsCsO4
Linear Formula CsH2AsO4
Formula Weight 273.84
MDL Number MFCD00143942
UN Number UN1557
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1764409

  SDS
alfa aesar™
A1764409
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1764418

  SDS
alfa aesar™
A1764418
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 273.84
Chemical Name or Material Cesium dihydrogen arsenate
Odor Odorless
Assay 99.99%
Molecular Formula H2AsCsO4
Linear Formula CsH2AsO4
MDL Number MFCD00143942
CAS 16331-85-4
UN Number UN1557
Quantity 10g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H331-H350
Toxic if swallowed.
Toxic if inhaled.
May cause cancer.
Health Hazard 3 GHS P Statement
P261-P281-P301+P310-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Use personal protective equipment as required.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ARSENIC COMPOUNDS, SOLID, N.O.S.
RTECS Number EV5790000
Safety and Handling

Danger

GHS P Statement
P261-P281-P301+P310-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Use personal protective equipment as required.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H331-H350
Toxic if swallowed.
Toxic if inhaled.
May cause cancer.