Please login to your online account to display your discounted pricing

Alfa Aesar™ Cesium dichromate, 99.8% (metals basis)

Chemical Identifiers

Chemical Name or Material Cesium dichromate
CAS 13530-67-1
Linear Formula Cs2Cr2O7
Formula Weight 481.8
MDL Number MFCD00049436
UN Number UN3087
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4023014

 
alfa aesar™
4023014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4023022

 
alfa aesar™
4023022
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 481.8
Physical Form Coarse Powder
Assay 99.8%
Sensitivity Hygroscopic
Assay Percent Notes (metals basis)
Linear Formula Cs2Cr2O7
CAS 13530-67-1
UN Number UN3087
Quantity 25g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H350-H272-H317
May cause cancer.
May intensify fire
oxidizer.
May cause an allergic skin reaction.
Health Hazard 3 GHS P Statement
P221-P210-P220-P363-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Keep/Store away from clothing/combustible materials.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: OXIDIZING SOLID, TOXIC, N.O.S.
Chemical Name or Material Cesium dichromate
MDL Number MFCD00049436
Safety and Handling

Danger

GHS P Statement
P221-P210-P220-P363-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Keep/Store away from clothing/combustible materials.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H350-H272-H317
May cause cancer.
May intensify fire
oxidizer.
May cause an allergic skin reaction.