Please login to your online account to display your discounted pricing

Alfa Aesar™ Cesium dichromate, 98%

Chemical Identifiers

Chemical Name or Material Cesium dichromate
CAS 13530-67-1
Molecular Formula Cs2Cr2O7
Formula Weight 481.8
MDL Number MFCD00049436
UN Number UN3087
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1856806

  SDS
alfa aesar™
A1856806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 481.8
Chemical Name or Material Cesium dichromate
Assay 98%
Molecular Formula Cs2Cr2O7
MDL Number MFCD00049436
CAS 13530-67-1
UN Number UN3087
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H350-H272-H317
May cause cancer.
May intensify fire; oxidizer.
May cause an allergic skin reaction.
Health Hazard 3 GHS P Statement
P221-P210-P220-P363-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Keep/Store away from clothing/combustible materials.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: OXIDIZING SOLID, TOXIC, N.O.S.
Quantity 5g
Safety and Handling

Danger

GHS P Statement
P221-P210-P220-P363-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Keep/Store away from clothing/combustible materials.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H350-H272-H317
May cause cancer.
May intensify fire; oxidizer.
May cause an allergic skin reaction.