missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Cesium chromate, 99.9% (metals basis)

Manufacturer:  Alfa Aesar™ 1349214

 View more versions of this product

Catalog No. AA1349214



Specifications

Specifications

Cesium chromate
(metals basis)
MFCD00016050
cesium chromate, cesium chromate 99.9+%, dicesium dioxido dioxo chromium, dicesium chromate, cesium chromate cs2cro4, chromic acid h2cro4 , dicesium salt, caesium chromate, cro4.2cs, acmc-1bym2, dicaesium 1+ chromate 2-
[O-][Cr](=O)(=O)[O-].[Cs+].[Cs+]
381.803
381.81
99.9%
Hygroscopic
13454-78-9
CrCs2O4
UN3087
BROHICCPQMHYFY-UHFFFAOYSA-N
dicesium;dioxido(dioxo)chromium
61613
Powder
Odorless
25g

Chemical Identifiers

13454-78-9
381.803
BROHICCPQMHYFY-UHFFFAOYSA-N
61613
[O-][Cr](=O)(=O)[O-].[Cs+].[Cs+]
CrCs2O4
MFCD00016050
cesium chromate, cesium chromate 99.9+%, dicesium dioxido dioxo chromium, dicesium chromate, cesium chromate cs2cro4, chromic acid h2cro4 , dicesium salt, caesium chromate, cro4.2cs, acmc-1bym2, dicaesium 1+ chromate 2-
dicesium;dioxido(dioxo)chromium
Safety and Handling

Safety and Handling

GHS H Statement
H350-H317
May cause cancer.
May cause an allergic skin reaction.

GHS P Statement
P261-P280-P281-P363-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: OXIDIZING SOLID, TOXIC, N.O.S.

TSCA : TSCA

Documents